Learn More
Greiner Bio-One 1536-well HiBase Polystyrene Microplates
Suited for luminescence measurements. Greiner Bio-One 1536-Well HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
4330.00€
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Description | Plate design: HiBase |
Array | 32 x 48 |
Material | Polystyrene |
Description
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- Sample carryover and cross-contamination risk is minimized
- Total
Certifications
DIN ISO 9001
Specifications
Standard | |
Plate design: HiBase | |
Polystyrene | |
Non-sterile |
Without Lid | |
32 x 48 | |
1536 | |
Flat Bottom (F-well) |