Learn More
Fisherbrand™ 18-Place Slide Staining Tray Set, Black
Slide tray that is ideal for clinical and research laboratory staining, rinsing, and drying applications.
Brand: Fisherbrand™ 120879F
140.60 EUR valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
- Durably constructed from solvent resistant polypropylene blend
- Non-skid silicone feet keep tray in place
- Two ways to empty stained liquid
- Equipped with handle cut outs for easy handling
- Humidity retention wells for specialized staining needs
Specifications
Slide Staining Tray | |
Microscope slide staining, rinsing and drying applications | |
1 black base and tray, 1 black cover, and 2 drain plugs |
18 | |
Standard microscope slides (76 x 26 mm) | |
1 Ea. |
Promotions
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now