missing translation for 'onlineSavingsMsg'
Learn More
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+% Product Code.: 15425466

(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%

Product Code. 15425466
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15425466

Brand: Thermo Scientific Alfa Aesar H60084.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452304-59-5 | C18H22NP | 283.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452304-59-5
Molecular Formula C18H22NP
Molecular Weight (g/mol) 283.36
MDL Number MFCD17013996
InChI Key ZATLZEHZPXYMFE-YTXKPMTANA-N
Synonym 1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min
PubChem CID 11166387
SMILES N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 43°C to 48°C
Flash Point >110°C (230°F)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Optical Rotation −79° (c=0.5 in Chloroform)
IUPAC Name 2-(diphenylphosphanyl)cyclohexan-1-amine
Formula Weight 283.35
Percent Purity ≥97%
Chemical Name or Material (1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD02Z006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60084.MD,H60084.03
Lot NumberU30F009 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60084.MD,H60084.03
Lot NumberR20F007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60084.MD,H60084.03
Lot NumberZ18D050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60084.MD,H60084.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.