Learn More
2,3,4,5,6-Pentafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals
Product Code. 11400063
Click to view available options
Quantity:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 440-60-8
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00004602
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
PubChem CID 9923
ChEBI CHEBI:44903
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O
Specifications
Melting Point 32°C to 36°C
Boiling Point 182°C
Flash Point 87°C (188°F)
Quantity 25 g
Beilstein 2052669
Formula Weight 198.09
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol