missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimethylbenzenesulfonic acid hydrate, 98% Product Code.: 11457237

2,5-Dimethylbenzenesulfonic acid hydrate, 98%

Product Code. 11457237
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11457237

Brand: Thermo Scientific Alfa Aesar A15007.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 609-54-1 | C8H10O3S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-54-1
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00151010
InChI Key IRLYGRLEBKCYPY-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
PubChem CID 11868
IUPAC Name 2,5-dimethylbenzenesulfonic acid
SMILES CC1=CC=C(C)C(=C1)S(O)(=O)=O

Specifications

Melting Point 84°C to 86°C
Quantity 25 g
Formula Weight 186.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylbenzenesulfonic acid hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238457 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15007.14,A15007.36,A15007.22
Lot Number10117936 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15007.14,A15007.36,A15007.22
Lot Number10235121 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15007.14,A15007.36,A15007.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethylbenzenesulfonic acid hydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.