missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals Product Code.: 11311357

2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals

Product Code. 11311357
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11311357

Brand: Thermo Scientific Alfa Aesar L01624.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

Solubility
Difficult in mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53957-33-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004625
InChI Key LEBQTCCCNMTXSF-UHFFFAOYSA-N
PubChem CID 94560
IUPAC Name (2,5-dimethylphenyl)methanol
SMILES CC1=CC(=C(C=C1)C)CO

Specifications

Melting Point 40°C to 42°C
Density 1.011
Boiling Point 232°C to 234°C
Flash Point 107°C (224°F)
Refractive Index 1.536
Quantity 5 g
Beilstein 2243420
Synonym 2,5-dimethylbenzyl alcohol, 2,5-dimethylphenyl methanol, benzenemethanol, 2,5-dimethyl, acmc-1ayay, 2,5-dimethylbenzylalcohol, benzenemethanol,2,5-dimethyl, 2,5-dimethylphenyl methanol #, 2'-oxy-1.2.4-trimethylbenzol, 2,5-dimethyl-hydroxymethylbenzene
Solubility Information Difficult in mix.
Formula Weight 136.19
Percent Purity ≥97%
Chemical Name or Material 2,5-Dimethylbenzyl alcohol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242447 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberL01624.06,L01624.03
Lot NumberH23Q26 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01624.06,L01624.03
Lot Number10112893 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01624.06,L01624.03
Lot Number10146045 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01624.06,L01624.03
Lot Number10223562 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01624.06,L01624.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.