missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimethylthiophene-3-boronic acid, 95% Product Code.: 15435277

2,5-Dimethylthiophene-3-boronic acid, 95%

Product Code. 15435277
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15435277

Brand: Thermo Scientific Alfa Aesar H32543.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 162607-23-0 | C6H9BO2S | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162607-23-0
Molecular Formula C6H9BO2S
Molecular Weight (g/mol) 156.01
MDL Number MFCD09837620
InChI Key RFQHMNWNEOZRLK-UHFFFAOYSA-N
Synonym 2,5-dimethylthiophene-3-boronic acid, 2,5-dimethylthiophen-3-yl boronic acid, 2,5-dimethyl-3-thienyl boronic acid, boronic acid,b-2,5-dimethyl-3-thienyl, 2,5-dimethyl-3-thienylboronic acid, 2,5-dimethylthien-3-ylboronic acid, boronic acid, 2,5-dimethyl-3-thienyl
PubChem CID 45588123
IUPAC Name (2,5-dimethylthiophen-3-yl)boronic acid
SMILES CC1=CC(B(O)O)=C(C)S1

Specifications

Quantity 1 g
Formula Weight 156.01
Percent Purity 95%
Chemical Name or Material 2,5-Dimethylthiophene-3-boronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247827 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog NumberH32543.MD,H32543.03
Lot Number10178639 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32543.MD,H32543.03
Lot Number10173430 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32543.MD,H32543.03
Lot NumberSP328-3 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32543.MD,H32543.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethylthiophene-3-boronic acid, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.