missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11434057
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19241-16-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00038715
InChI Key UULUECCNPPJFBU-UHFFFAOYSA-N
PubChem CID 87976
IUPAC Name 2-isothiocyanato-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)N=C=S

Specifications

Density 1.085
Boiling Point 247°C
Quantity 25 g
Odor Pungent
Refractive Index 1.625
UN Number UN2922
Beilstein 637195
Synonym 2,6-dimethylphenyl isothiocyanate, 2,6-xylyl isothiocyanate, 2,6-dimethylphenylisothiocyanate, benzene, 2-isothiocyanato-1,3-dimethyl, 2-isothiocyanato-1,3-dimethyl-benzene, benzene,2-isothiocyanato-1,3-dimethyl, 2,6-dimethylbenzenisothiocyanate, acmc-20ao7z, 2,6-dimethylphenyl-isothiocyanate, 2,6-dimethylphenyl-1-isothiocyanate
Solubility Information Hydrolyzes in water.
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'