Learn More
2-Amino-5-bromothiazole monohydrobromide, 95% Product Code.: 10754451
2-Amino-5-bromothiazole monohydrobromide, 95%
Product Code. 10754451 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Product Code. 10754451
Brand: Thermo Scientific Acros 156000250
Request Bulk or Custom Format
CAS: 61296-22-8 | C3H3BrN2S·HBr | 259.95 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 61296-22-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point 165°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H3BrN2S·HBr
MDL Number MFCD00012712
Quantity 25 g
Synonym 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7
InChI Key NUSVDASTCPBUIP-UHFFFAOYSA-N
SMILES C1=C(SC(=N1)N)Br.Br
IUPAC Name 5-bromo-1,3-thiazol-2-amine;hydrobromide
Molecular Weight (g/mol) 259.95
PubChem CID 2723848
Formula Weight 259.95
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromothiazole monohydrobromide, 95%
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0470059 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number156000250
Lot NumberA0388303 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number156000250
Product Identifier
  • 2-Amino-5-bromothiazole monohydrobromide
Product Title
2-Amino-5-bromothiazole monohydrobromide, 95% >