missing translation for 'onlineSavingsMsg'
Learn More
2-(Boc-amino)ethanol, 95%, Thermo Scientific Chemicals Product Code.: 15425077

2-(Boc-amino)ethanol, 95%, Thermo Scientific Chemicals

Product Code. 15425077
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15425077

Brand: Thermo Scientific Alfa Aesar H32356.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26690-80-2 | C7H15NO3 | 161.201 g/mol

Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26690-80-2
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00056657
InChI Key GPTXCAZYUMDUMN-UHFFFAOYSA-N
Synonym n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl esterShow MoreShow Less
PubChem CID 2733206
IUPAC Name tert-butyl N-(2-hydroxyethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCO

Specifications

Boiling Point 92°C (0.22mmHg)
Linear Formula (CH3)3CCO2NHCH2CH2OH
Refractive Index 1.449
Solubility Information Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.
Density 1.042
Quantity 5 g
Formula Weight 161.2
Percent Purity 95%
Chemical Name or Material 2-(Boc-amino)ethanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10202742 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberH32356.14,H32356.06
Lot Number10241664 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberH32356.14,H32356.06
Lot Number10182497 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32356.14,H32356.06
Lot Number5039A12W Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32356.14,H32356.06
Lot Number10172225 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32356.14,H32356.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Boc-amino)ethanol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.