missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4-chloro-1-fluorobenzene, 97% Product Code.: 11425700

2-Bromo-4-chloro-1-fluorobenzene, 97%

Product Code. 11425700
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11425700

Brand: Thermo Scientific Alfa Aesar H25935.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1996-30-1 | C6H3BrClF | 209.442 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210Show MoreShow Less
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F

Specifications

Density 1.74
Boiling Point 180°C
Flash Point 85°C (185°F)
Refractive Index 1.554
Quantity 25 g
Formula Weight 209.45
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-chloro-1-fluorobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10157232 Certificate TypeCertificate of Analysis Date05/05/2023 Product CodeH25935.06,H25935.14
Lot Number10144466 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH25935.06,H25935.14
Lot NumberE15T048 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH25935.06,H25935.14
Lot NumberA26W014 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH25935.06,H25935.14
Lot Number10157232 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH25935.06,H25935.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4-chloro-1-fluorobenzene, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.