missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-5-methyl-1-benzothiophene, 97%, Thermo Scientific™ Product Code.: 10536314

2-Bromo-5-methyl-1-benzothiophene, 97%, Thermo Scientific™

Product Code. 10536314
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10536314

Brand: Thermo Scientific Maybridge MO07070DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 111860-00-5
Molecular Formula C9H7BrS
Molecular Weight (g/mol) 227.119
MDL Number MFCD05865140
InChI Key HZPBPLLBZYSGBI-UHFFFAOYSA-N
Synonym 2-bromo-5-methylbenzo b thiophene, benzo b thiophene,2-bromo-5-methyl, acmc-20mexj, 2-bromanyl-5-methyl-1-benzothiophene, 2-bromo-5-methyl-1-benzo b thiophene, benzo b thiophene, 2-bromo-5-methylShow MoreShow Less
PubChem CID 2794769
IUPAC Name 2-bromo-5-methyl-1-benzothiophene
SMILES CC1=CC2=C(C=C1)SC(=C2)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 227.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methyl-1-benzothiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-5-methyl-1-benzothiophene, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.