missing translation for 'onlineSavingsMsg'
Learn More
2-(Carboxymethylthio)succinic acid, 97%, Thermo Scientific™ Product Code.: 15433197

2-(Carboxymethylthio)succinic acid, 97%, Thermo Scientific™

Product Code. 15433197
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15433197

Brand: Thermo Scientific Alfa Aesar A19818.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-68-3
Molecular Formula C6H8O6S
Molecular Weight (g/mol) 208.184
MDL Number MFCD00002720
InChI Key JPHVSDWIWBDHOC-UHFFFAOYSA-N
Synonym carboxymethylmercaptosuccinic acid, butanedioic acid, carboxymethyl thio, carboxymethylthio succinic acid, 2-carboxymethyl sulfanyl butanedioic acid, 2-carboxymethyl thio succinic acid, carboxymethyl thio succinic acid, 2-carboxymethylsulfanyl butanedioic acid, 2-carboxymethyl sulfanyl succinic acid, butanedioic acid, 2-carboxymethyl thio
PubChem CID 97922
IUPAC Name 2-(carboxymethylsulfanyl)butanedioic acid
SMILES C(C(C(=O)O)SCC(=O)O)C(=O)O

Specifications

Melting Point 138°C to 141°C
Quantity 10 g
Formula Weight 208.19
Percent Purity 97%
Chemical Name or Material 2-(Carboxymethylthio)succinic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC7349A Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Carboxymethylthio)succinic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.