missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15491688
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-06-7
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD00061094
InChI Key WLXRKCGYQAKHSJ-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl benzoicacid, 2-chloro-5-trifluoromethyl-benzoic acid, maybridge1_003457, pubchem1377, pubchem7879, acmc-209xwb, 2-chloro5-trifluoromethylbenzoic acid
PubChem CID 688182
IUPAC Name 2-chloro-5-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl

Specifications

Melting Point 93°C to 96°C
Quantity 5 g
Formula Weight 224.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'