missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-6-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals Product Code.: 11445367

2-Chloro-6-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Product Code. 11445367
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11445367

Brand: Thermo Scientific Alfa Aesar B22714.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-94-3
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00272565
InChI Key OTRRSPQJZRCMDA-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl aniline, 2-amino-3-chlorobenzotrifluoride, benzenamine, 2-chloro-6-trifluoromethyl, 2-chloranyl-6-trifluoromethyl aniline, pubchem2764, 2-chloro-6-trifluoromethyl benzenamine, 2-chloro-6-trifluoromethyl phenylamine, 6-chloro-2-trifluoromethyl phenylamine, 2-chloro-6-trifluoromethyl-benzenamine, 2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluorideShow MoreShow Less
PubChem CID 2756384
IUPAC Name 2-chloro-6-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=CC=C1C(F)(F)F

Specifications

Density 1.42
Boiling Point 39°C to 40°C (0.1 mmHg)
Quantity 5 g
UN Number UN2810
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10195691 Certificate TypeCertificate of Analysis Date23/06/2023 Catalog NumberB22714.06,B22714.14,B22714
Lot Number10127982 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22714.06,B22714.14
Lot NumberI2676A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22714.06,B22714.14
Lot Number10195691 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22714.06,B22714.14
Lot Number10143434 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22714.06,B22714.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-6-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.