missing translation for 'onlineSavingsMsg'
Learn More
2-Cyclopenten-1-one, 98%, Thermo Scientific Chemicals Product Code.: 11481017

2-Cyclopenten-1-one, 98%, Thermo Scientific Chemicals

Product Code. 11481017
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11481017

Brand: Thermo Scientific Alfa Aesar B23408.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-30-3 | C5H6O | 82.10 g/mol

2-Cyclopenten-1-one is a versatile electrophile employed in various addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with silyl enol ethers and siloxanes. It is also used in Diels-Alder cycloadditions and phosphoniosilylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopenten-1-one is a versatile electrophile employed in various addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with silyl enol ethers and siloxanes. It is also used in Diels-Alder cycloadditions and phosphoniosilylations.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-30-3
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.10
MDL Number MFCD00001401
InChI Key BZKFMUIJRXWWQK-UHFFFAOYSA-N
Synonym 2-cyclopenten-1-one, 2-cyclopentenone, cyclopentenone, cyclopent-2-enone, cyclopenten-3-one, 2-cyclopentene-1-one, cyclopent-2-ene-1-one, unii-q0u2igf9ck, 3-cyclopenten-2-one, q0u2igf9ckShow MoreShow Less
PubChem CID 13588
IUPAC Name cyclopent-2-en-1-one
SMILES O=C1CCC=C1

Specifications

Density 1.01
Boiling Point 150°C
Flash Point 42°C (108°F)
Odor Characteristic
Refractive Index 1.481
Quantity 25 g
UN Number UN1224
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 82.1
Percent Purity 98%
Chemical Name or Material 2-Cyclopenten-1-one
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU14K034 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberB23408.06,B23408.14
Lot NumberX13J034 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberB23408.06,B23408.14
Lot Number10236646 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog NumberB23408.06,B23408.14
Lot Number10232467 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog NumberB23408.06,B23408.14
Lot NumberY22I040 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberB23408.06,B23408.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Cyclopenten-1-one, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.