missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™ Product Code.: 15427615

2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15427615
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15427615

Brand: Thermo Scientific Alfa Aesar H26714.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As intermediates. Commonly used laboratory reagent. Used in the field of nanoparticles.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-98-9
Molecular Formula C9H6F4O3
Molecular Weight (g/mol) 238.14
MDL Number MFCD04115892
InChI Key PYCWSPHSXHTRQT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethoxy phenylacetic acid, 2-2-fluoro-5-trifluoromethoxy phenyl acetic acid, 2-fluoro-5-trifluoromethoxy phenyl acetic acidShow MoreShow Less
PubChem CID 2783328
IUPAC Name 2-[2-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
SMILES OC(=O)CC1=C(F)C=CC(OC(F)(F)F)=C1

Specifications

Melting Point 54°C to 56°C
Quantity 250 mg
Formula Weight 238.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)phenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10202945 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10156675 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10141193 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10164735 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.