missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11324556
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89787-12-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD03411937
InChI Key KTZUVUWIBZMHMC-UHFFFAOYSA-N
Synonym 2-isopropylphenylboronic acid, 2-isopropylphenyl boronic acid, 2-isopropylbenzeneboronic acid, 2-cumylboronic acid, 2-propan-2-ylphenyl boronic acid, o-isopropylphenylboronic acid, 2-isopropylphenboronic acid, 2-isopropylphenyboronic acid, 2-propan-2-yl phenyl boronic acid, boronic acid, 2-1-methylethyl phenyl
PubChem CID 2773477
IUPAC Name (2-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC=C1B(O)O

Specifications

Melting Point 85°C to 86°C
Quantity 250 mg
Beilstein 3090298
Formula Weight 164.01
Percent Purity 97%
Chemical Name or Material 2-Isopropylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'