missing translation for 'onlineSavingsMsg'
Learn More
2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals Product Code.: 11453494

2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11453494
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11453494

Brand: Thermo Scientific Alfa Aesar A11192.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27231-36-3
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00010617
InChI Key CWIYBOJLSWJGKV-UHFFFAOYSA-N
Synonym 2-mercapto-5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole-2 3h-thione, 5-methyl-1h-benzimidazole-2-thiol, 5-methyl-2-benzimidazolethiol, 5-methyl-2-mercaptobenzimidazole, 5-methyl-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione, 1,3-dihydro-5-methyl, 1,3-dihydro-5-methyl-2h-benzimidazole-2-thione, 5-methyl-1h-benzoimidazole-2-thiol, 5-methylbenzimidazole-2-thiolShow MoreShow Less
PubChem CID 712373
IUPAC Name 5-methyl-1,3-dihydrobenzimidazole-2-thione
SMILES CC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point ∼295°C (decomposition)
Quantity 25 g
Beilstein 5083
Solubility Information Insoluble in water.
Formula Weight 164.23
Percent Purity 98%
Chemical Name or Material 2-Mercapto-5-methylbenzimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253496 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberA11192.22,A11192.06,A11192.14
Lot Number10245461 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog NumberA11192.22,A11192.06,A11192.14
Lot Number10048118 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11192.22,A11192.06,A11192.14
Lot Number10231211 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11192.22,A11192.06,A11192.14
Lot Number10212721 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11192.22,A11192.06,A11192.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.