Learn More
2-Mercaptobenzothiazole, 95%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 172.0°C to 182.0°C |
Color | Yellow |
Flash Point | 243°C |
Loss on Drying | 0.5% max. (60°C, 3 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Plastic bottle |
Quantity | 50 g |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling


- 2-Mercaptobenzothiazole
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-Mercaptobenzothiazole
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.