missing translation for 'onlineSavingsMsg'
Learn More
2-Mercaptoethanol, 98+%, Thermo Scientific Chemicals Product Code.: 11442608

2-Mercaptoethanol, 98+%, Thermo Scientific Chemicals

Product Code. 11442608
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11442608

Brand: Thermo Scientific Alfa Aesar A15890.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Mercaptoethanol, CAS # 60-24-2, is commonly used as a reducing agent in laboratories and has a variety of applications such as solubilizing proteins.

Solubilizes proteins by reducing disulfide linkages2-Mercaptoethanol is used in the preparation of nano-graphene for cellular imaging and drug delivery as well as multifunctional polymeric micelle. It acts as a reducing agent used in electrophoresis, amino acid detection, and distinguishing ssDNA/dsDNA. It is also employed as a standard buffer. It is also used in the preparation of PVC heat stabilizers and as a chain transfer agent in the manufacture of PVC. Further, it is used in some RNA isolation procedures to eliminate ribonuclease. It is utilized as a corrosion inhibitor and ore floatation agent. In biochemistry, it is useful to study the activity of the immune system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 2-Mercaptoethanol is a reducing agent that can be used in studies to solubilize proteins by reducing disulfide linkages

Application

• 2-Mercaptoethanol acts as a reducing agent in electrophoresis, amino acid detection, and distinguishing ssDNA/dsDNA
• It can also be used as a standard buffer
• It can also be used in some RNA isolation procedures to eliminate ribonuclease
• In biochemistry, it is useful for studying the activity of the immune system
• Furthermore, it can be used in laboratory applications involving preparation of multifunctional polymeric micelles and nano-graphene for cellular imaging

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Melting Point -100°C
Density 1.114
Boiling Point 156°C to 158°C
Flash Point 73°C (163°F)
Odor Unpleasant
Refractive Index 1.5
Quantity 1000 g
UN Number UN2966
Beilstein 773648
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,5869
Solubility Information Miscible with water,alcohol,ether,benzene and other organic solvents.
IUPAC Name 2-sulfanylethan-1-ol
Formula Weight 78.13
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Mercaptoethanol, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.