Learn More
2-Methoxy-3-(trifluoromethyl)aniline, 97%, Thermo Scientific™
102.00€ - 407.00€
Chemical Identifiers
CAS | 634187-08-9 |
---|---|
Molecular Formula | C8H8F3NO |
Molecular Weight (g/mol) | 191.153 |
MDL Number | MFCD08532489 |
InChI Key | QOMZMKOUJOLVKC-UHFFFAOYSA-N |
Synonym | 2-methoxy-3-trifluoromethyl aniline, 3-trifluoromethyl-o-anisidine, 3-amino-2-methoxybenzotrifluoride, 2-amino-6-trifluoromethyl anisole, 2-methoxy-3-trifluoromethyl benzenamine, 2-methoxy-3-trifluoromethyl phenylamine |
PubChem CID | 20112025 |
IUPAC Name | 2-methoxy-3-(trifluoromethyl)aniline |
SMILES | COC1=C(C=CC=C1N)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilityNot miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
634187-08-9 | |
191.153 | |
QOMZMKOUJOLVKC-UHFFFAOYSA-N | |
20112025 | |
COC1=C(C=CC=C1N)C(F)(F)F |
C8H8F3NO | |
MFCD08532489 | |
2-methoxy-3-trifluoromethyl aniline, 3-trifluoromethyl-o-anisidine, 3-amino-2-methoxybenzotrifluoride, 2-amino-6-trifluoromethyl anisole, 2-methoxy-3-trifluoromethyl benzenamine, 2-methoxy-3-trifluoromethyl phenylamine | |
2-methoxy-3-(trifluoromethyl)aniline |
Specifications
634187-08-9 | |
MFCD08532489 | |
2-methoxy-3-trifluoromethyl aniline, 3-trifluoromethyl-o-anisidine, 3-amino-2-methoxybenzotrifluoride, 2-amino-6-trifluoromethyl anisole, 2-methoxy-3-trifluoromethyl benzenamine, 2-methoxy-3-trifluoromethyl phenylamine | |
QOMZMKOUJOLVKC-UHFFFAOYSA-N | |
2-methoxy-3-(trifluoromethyl)aniline | |
20112025 | |
97% |
C8H8F3NO | |
UN2810 | |
Not miscible or difficult to mix. | |
COC1=C(C=CC=C1N)C(F)(F)F | |
191.153 | |
191.15 | |
2-Methoxy-3-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Recommended Storage : Ambient temperatures