missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-8-nitroquinoline, 98%, Thermo Scientific Chemicals

Product Code. 11317935
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881-07-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00006764
InChI Key UHPGVDHXHDPYQP-UHFFFAOYSA-N
PubChem CID 13433
IUPAC Name 2-methyl-8-nitroquinoline
SMILES CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O

Specifications

Melting Point 136°C to 139°C
Odor Odorless
Quantity 1 g
Beilstein 145077
Synonym 8-nitroquinaldine, quinaldine, 8-nitro, quinoline, 2-methyl-8-nitro, unii-xrv949auqz, xrv949auqz, acmc-209qr6, 2-methyl-8-nitro-quinoline, 5-20-07-00387 beilstein handbook reference, quinoline,2-methyl-8-nitro, uhpgvdhxhdpyqp-uhfffaoysa
Formula Weight 188.19
Percent Purity 98%
Chemical Name or Material 2-Methyl-8-nitroquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'