Learn More
2-Nitrobenzylamine hydrochloride, 98%
Description
2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 250°C (decomposition) |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Formula Weight | 188.62 |
Percent Purity | 98% |
Chemical Name or Material | 2-Nitrobenzylamine hydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.