missing translation for 'onlineSavingsMsg'
Learn More
2-Pentynoic acid, 97%, Thermo Scientific Chemicals Product Code.: 15400198

2-Pentynoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15400198
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15400198

Brand: Thermo Scientific Alfa Aesar H53411.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5963-77-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00014370
InChI Key MINRDQDGBLQBGD-UHFFFAOYSA-N
Synonym 2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
PubChem CID 324379
IUPAC Name pent-2-ynoic acid
SMILES CCC#CC(=O)O

Specifications

Melting Point 47°C to 53°C
Flash Point 96°C (205°F)
Linear Formula CH3CH2CH≡CCO2H
Quantity 5 g
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Pentynoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP28I029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH53411.03,H53411.14,H53411.06
Lot NumberM19E063 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53411.03,H53411.14,H53411.06
Lot NumberW25E041 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53411.03,H53411.14,H53411.06
Lot NumberT17D020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53411.03,H53411.14,H53411.06
Lot NumberL21X016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53411.03,H53411.14,H53411.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Pentynoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.