missing translation for 'onlineSavingsMsg'
Learn More
2-tert-Butylaniline, 98%, Thermo Scientific Chemicals Product Code.: 10110622

2-tert-Butylaniline, 98%, Thermo Scientific Chemicals

Product Code. 10110622
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10110622

Brand: Thermo Scientific Acros 369870250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6310-21-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point -60°C
Density 0.9570g/mL
Boiling Point 123°C to 124°C (17.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H15N
Linear Formula (CH3)3CC6H4NH2
Refractive Index 1.5440 to 1.546
MDL Number MFCD00130023
Quantity 25 g
Specific Gravity 0.957
Synonym 2-tert-butyl aniline, 2-tert-butylphenylamine, benzenamine, 2-1,1-dimethylethyl, 2-t-butylaniline, 2-tert-butyl-phenylamine, 2-1,1-dimethylethyl benzenamine, 2-tert-butyl phenylamine, pubchem20188, aniline, 2-tert-butyl
InChI Key AEIOZWYBDBVCGW-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC=CC=C1N
IUPAC Name 2-tert-butylaniline
Molecular Weight (g/mol) 149.24
PubChem CID 80574
Formula Weight 149.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-tert-Butylaniline, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467942 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number369870250
Lot NumberA0460353 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number369870250
Lot NumberA0429240 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog Number369870250
Lot NumberA0421163 Certificate TypeCertificate of Analysis Date04/08/2020 Catalog Number369870250
Lot NumberA0391624 Certificate TypeCertificate of Analysis Date10/06/2020 Catalog Number369870250
compliance-icons
Product Identifier
  • 2-tert-Butylaniline
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-tert-Butylaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.