Learn More
uvex 2111 Clear Earplug Box
Pocket-sized ear plug case protects GHS plugs of all kinds from damage, and contamination from unhygienic storage.
Brand: uvex 2111404
32.05 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
- Pocket sized ear plug case
- Protects GHS ear plugs from damage
- Prevents contamination from unhygienic storage
Specifications
Transparent | |
Case | |
Reusable |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08