Learn More
uvex 2112 X-Fit Disposable Ear Plugs
Ergonomic, pre-shaped ear plugs are characterized by very high insulation values making them suitable for use in extremely noisy environments.
Brand: uvex 2112094
This item is not returnable.
View return policy
Description
- Ergonomically pre-shaped earplugs characterized by very high insulation values
- Soft foam offers a high level of comfort, even after long periods of use
- Suitable for use in extremely noisy environments
- Available in a hygienic polybag, or retail minibox
Specifications
Lime Green | |
Regular | |
Polybag | |
37 dB | |
Oval | |
Single-use | |
No |
No | |
2112 X-Fit | |
100 | |
Uncorded | |
Ear Plugs | |
No | |
EN 352-2 |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
uvex 2112 X-Fit Disposable Ear Plugs > Polybag