Learn More
uvex 2112 Xtra-Fit Disposable Ear Plugs
Ergonomic, pre-formed ear plugs expand slowly to adapt to the size and shape of the ear canal, thus minimizing leakage and ensuring optimal protection.
21.10€ - 58.90€
Specifications
Color | Orange |
---|---|
ANSI S3.1974 | No |
Size | Large |
Model | 2112 Xtra-Fit |
Noise Reduction Rating (NRR) | 36 dB |
Product Code | Brand | Packaging Type | Pairs per Package | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Pairs per Package | Price | Quantity & Availability | |||||
17230342
|
uvex
2112093 |
Polybag | 100 |
21.10€
100 pairs |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
17210342
|
uvex
2112060 |
Polybag | 200 |
42.20€
200 pairs |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
17220342
|
uvex
2112061 |
Refill Box | 250 |
58.90€
250 pairs |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
Description
- Ergonomically, pre-shaped disposable earplugs
- Slowly expand to adapt to the size and shape of the ear canal
- Minimized leakage for optimal protection
- Suitable for larger ear canals
- Available in hygienic polybags, or refill bags
Specifications
Orange | |
Large | |
36 dB | |
Oval | |
Single-use | |
No |
No | |
2112 Xtra-Fit | |
Uncorded | |
Ear Plugs | |
No | |
EN 352-2 |
Product Title
uvex 2112 Xtra-Fit Disposable Ear Plugs