missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ 25 mM MgCl2
25 mM MgCl2 can be used for optimization of magnesium ion concentration in PCR reaction.
Specifications
Content And Storage | Store at -20°C |
---|---|
For Use With (Application) | Standard PCR |