Learn More
Thermo Scientific Alfa Aesar 2500ML Pyridine, HPLC Grade, 99.5+%
Brand:  Thermo Scientific Alfa Aesar 022905.0F
2500ML Pyridine, HPLC Grade, 99.5+%
Product Code. 15481097
  • POA / 2500mL
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY30K631 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog Number022905.K7,022905.M1,022905.0F,022905.K2
Lot NumberX28K963 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number022905.K7,022905.M1,022905.0F,022905.K2
Lot NumberX04K962 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number022905.K7,022905.M1,022905.0F,022905.K2
Lot NumberX04K980 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number022905.K7,022905.M1,022905.0F,022905.K2
Lot NumberT30K879 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number022905.K7,022905.M1,022905.0F,022905.K2
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Alfa Aesar 2500ML Pyridine, HPLC Grade, 99.5+% >