Learn More
Fisher Chemical 25KG CALCIUM CHLORIDE ANALYSIS EP&USPSPEC.
Brand:  Fisher Chemical C/1500/70
25KG CALCIUM CHLORIDE ANALYSIS EP&USPSPEC.
Product Code. 11906961
  • ONLINE EXCLUSIVE PRICE
    617.00€
    599.63€ / 25kg
    Save 17.37€
    2% Off
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.