Learn More
Testosterone Propionate EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Testosterone Propionate EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate) |
| CAS | 1169-49-9 |
| Analyte or Component Names | 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H34 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Oxoandrost-4-en-17β-yl 2-Methyl-propanoate (Testosterone Isobutyrate), Testosterone Isocaproate Imp. D (EP), Testosterone Propionate Imp. B (EP), 3-Oxoandrost-4-en-17beta-yl 2-Methylpropanoate, Testosterone isobutyrate, Androst-4-en-3-one, 17-(2-methyl-1-oxopropoxy)-, (17beta)-, Testosterone, isobutyrate (6CI,7CI,8CI), (17beta)-17-(2-Methyl-1-oxopropoxy)androst-4-en-3-one, Isobutyric acid, ester with testosterone (8CI), 17beta-(Isobutyroxy)androst-4-en-3-one, 3-Oxoandrost-4-en-17beta-yl isobutyrate, Agovirin-Depot, Perandren M, Testocryst, Testosterone 17-isobutyrate, Testosterone 2-methylpropanoate, Testosterone 2-methylpropionate, Testosterone Isocaproate Impurity D, Testosterone Propionate Impurity B |
| InChI Formula | InChI=1S/C23H34O3/c1-14(2)21(25)26-20-8-7-18-17-6-5-15-13-16(24)9-11-22(15,3)19(17)10-12-23(18,20)4/h13-14,17-20H,5-12H2,1-4H3/t17-,18-,19-,20-,22-,23-/m0/s1 |
| SMILES | CC(C)C(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 2-methylpropanoate |
| Show More |