missing translation for 'onlineSavingsMsg'
Learn More

Propyl (2E)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate, Mikromol™

Product Code. 30049403
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049403

Brand: Mikromol MM1221.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Propyl (2E)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propyl (2E)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate
CAS 1364322-42-8
Analyte or Component Names Propyl (2E)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate
Shipping Condition Room Temperature
Molecular Formula C13 H12 N2 O6
Certifications/Compliance ISO 17025
Synonym Entacapone Imp. I (EP), Propyl (2E)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate, Entacapone Impurity I
InChI Formula InChI=1S/C14H21NO3.ClH/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11;/h6-9,13H,5,10H2,1-4H3;1H
SMILES CCCOC(=O)\C(=C\c1cc(O)c(O)c(c1)[N+](=O)[O-])\C#N
IUPAC Name propyl (E)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol) 292.24
Quantity 25 mg
Formula Weight 292.0695 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.