missing translation for 'onlineSavingsMsg'
Learn More
3-(2-Methyl-3-furylthio)-4-heptanone, 97%, Thermo Scientific™ Product Code.: 11337169

3-(2-Methyl-3-furylthio)-4-heptanone, 97%, Thermo Scientific™

Product Code. 11337169
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11337169

Brand: Thermo Scientific Alfa Aesar H27378.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61295-41-8
Molecular Formula C12H18O2S
Molecular Weight (g/mol) 226.33
MDL Number MFCD01723735
InChI Key GFRRQSASJZMMJC-UHFFFAOYNA-N
Synonym 3-2-methyl-3-furylthio-4-heptanone, 3-2-methylfuran-3-yl thio heptan-4-one, fema no. 3570, 3-2-methyl-3-furyl thio-4-heptanone, 1,3-diethylacetonyl 3-methyl-3-furyl sulfide, 3-2-methyl-3-furyl thio heptan-4-one, 4-heptanone, 3-2-methyl-3-furanyl thio, 4-heptanone, 3-2-methyl-3-furyl thio, 3-2-methylfuran-3-yl sulfanylheptan-4-one, 3-2-methyl-3-furanyl thio-4-heptanoneShow MoreShow Less
PubChem CID 43598
IUPAC Name 3-(2-methylfuran-3-yl)sulfanylheptan-4-one
SMILES CCCC(=O)C(CC)SC1=C(C)OC=C1

Specifications

Refractive Index 1.5015
Quantity 1 g
Formula Weight 226.34
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-3-furylthio)-4-heptanone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10133908 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Methyl-3-furylthio)-4-heptanone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.