missing translation for 'onlineSavingsMsg'
Learn More
3-(2-Thienyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™ Product Code.: 10613233

3-(2-Thienyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10613233
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10613233

Brand: Thermo Scientific Maybridge SP01369DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26033-27-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6N2OS
MDL Number MFCD02677688
Quantity 1 g
Synonym 3-thiophen-2-yl-1h-pyrazole-4-carbaldehyde, 3-2-thienyl-1h-pyrazole-4-carbaldehyde, 3-thien-2-yl-1h-pyrazole-4-carbaldehyde, 5-thiophen-2-yl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-2-thienyl, 3-thiophen-2-yl-2h-pyrazole-4-carbaldehyde, 3-2-thienyl pyrazole-4-carbaldehyde, 5-2-thienyl pyrazole-4-carbaldehyde, 3-2-thienyl-1h-pyrazole-4-carboxaldehyde
InChI Key DDQIKYLVCQKLHT-UHFFFAOYSA-N
SMILES C1=CSC(=C1)C2=C(C=NN2)C=O
IUPAC Name 5-thiophen-2-yl-1H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol) 178.209
PubChem CID 737231
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 3-(2-Thienyl)-1H-pyrazole-4-carbaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Thienyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.