missing translation for 'onlineSavingsMsg'
Learn More
3,3',5,5'-Tetramethylbiphenyl, 97+%, Thermo Scientific Chemicals Product Code.: 11307266

3,3',5,5'-Tetramethylbiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 11307266
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11307266

Brand: Thermo Scientific Alfa Aesar L09172.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,3',5,5'-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3',5,5'-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3',5,5'-tetracarboxylic acid by the oxidation of with potassium permanganate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25570-02-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00051910
InChI Key CMZYGFLOKOQMKF-UHFFFAOYSA-N
PubChem CID 520212
IUPAC Name 1-(3,5-dimethylphenyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Melting Point 47°C to 49°C
Quantity 5 g
Beilstein 1935931
Synonym 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl #
Formula Weight 210.32
Percent Purity ≥97%
Chemical Name or Material 3,3',5,5'-Tetramethylbiphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10179102 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberL09172.03,L09172.06
Lot NumberK16W037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL09172.03,L09172.06
Lot Number10179102 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL09172.03,L09172.06
Lot Number10163211 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL09172.03,L09172.06
Lot NumberI26Q017 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL09172.03,L09172.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,3',5,5'-Tetramethylbiphenyl, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.