missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trichloropyridine, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
Melting Point | 72.0°C to 73.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 182.44 |
Percent Purity | 98% |
Chemical Name or Material | 3,4,5-Trichloropyridine |
Safety and Handling
Product Identifier
- 3,4,5-Trichloropyridine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3,4,5-Trichloropyridine, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction