missing translation for 'onlineSavingsMsg'
Learn More
3,5-Difluorobenzyl bromide, 98%, Thermo Scientific™ Product Code.: 11428577

3,5-Difluorobenzyl bromide, 98%, Thermo Scientific™

Product Code. 11428577
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11428577

Brand: Thermo Scientific Alfa Aesar A18628.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141776-91-2
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00010304
InChI Key KVSVNRFSKRFPIL-UHFFFAOYSA-N
Synonym 3,5-difluorobenzyl bromide, 1-bromomethyl-3,5-difluorobenzene, 3,5-difluorobenzylbromide, alpha-bromo-3,5-difluorotoluene, 3,5-difluoro benzyl bromide, 3,5-bifluorobenzyl bromide, 1-bromomethyl-3,5-difluoro-benzene, benzene, 1-bromomethyl-3,5-difluoro, 5-bromomethyl-1,3-difluorobenzene, pubchem2245
PubChem CID 518896
IUPAC Name 1-(bromomethyl)-3,5-difluorobenzene
SMILES FC1=CC(CBr)=CC(F)=C1

Specifications

Density 1.6
Boiling Point 64°C to 65°C (4.5 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.521
Quantity 25 g
UN Number UN1760
Beilstein 7422062
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 3,5-Difluorobenzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10138723 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberA18628.06,A18628.03,A18628.14
Lot Number10186540 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberA18628.06,A18628.03,A18628.14
Lot Number10196465 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberA18628.06,A18628.03,A18628.14
Lot Number10207378 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberA18628.06,A18628.03,A18628.14
Lot Number10163987 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberA18628.06,A18628.03,A18628.14
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Difluorobenzyl bromide, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.