missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals Product Code.: 11470553

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11470553
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11470553

Brand: Thermo Scientific Alfa Aesar A12674.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 2458-12-0 | C8H9NO2 | 151.165 g/mol

3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2458-12-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 164°C to 168°C
Quantity 25 g
Beilstein 2082449
Solubility Information Very soluble in water.
Formula Weight 151.16
Percent Purity 99%
Chemical Name or Material 3-Amino-4-methylbenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10190116 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12674.06,A12674.14,A12674.22
Lot Number10148679 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12674.06,A12674.14,A12674.22
Lot NumberY06E041 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12674.06,A12674.14,A12674.22
Lot NumberX30E022 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12674.06,A12674.14,A12674.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.