Learn More
3-Amino-5-(2-furyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals
Product Code. 11362587
Click to view available options
Quantity:
250 mg
1 g
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96799-02-9
Molecular Formula C7H7N3O
Molecular Weight (g/mol) 149.153
MDL Number MFCD00082696
InChI Key XJNZHICOWTVWOX-UHFFFAOYSA-N
Synonym 3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine
PubChem CID 523183
IUPAC Name 5-(furan-2-yl)-1H-pyrazol-3-amine
SMILES C1=COC(=C1)C2=CC(=NN2)N
Specifications
Melting Point 124°C to 128°C
Quantity 1 g
Beilstein 4399373
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(2-furyl)-1H-pyrazole