missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-5-chloropyridine, 95%, Thermo Scientific™ Product Code.: 15474477

3-Amino-5-chloropyridine, 95%, Thermo Scientific™

Product Code. 15474477
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15474477

Brand: Thermo Scientific Alfa Aesar H31833.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22353-34-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD03701386
InChI Key ZHMASVAJFJFFLS-UHFFFAOYSA-N
PubChem CID 818257
IUPAC Name 5-chloropyridin-3-amine
SMILES C1=C(C=NC=C1Cl)N

Specifications

Melting Point 71°C to 75°C
Odor Unpleasant
Quantity 250 mg
Synonym 3-amino-5-chloropyridine, 5-chloro-3-pyridinamine, 5-chloro-pyridin-3-ylamine, 3-pyridinamine, 5-chloro, 5-chloro-3-pyridylamine, 5-chloro-3-aminopyridine, 5-chloropyridine-3-amine, 3-pyridinamine,5-chloro, 5-chloro-3-pyridinylamine, 3-amino-5-chloro-pyridine
Formula Weight 128.56
Percent Purity 95%
Chemical Name or Material 3-Amino-5-chloropyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10154563 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10163572 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10168512 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-5-chloropyridine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.