missing translation for 'onlineSavingsMsg'
Learn More

3-Boc-3-(methylamino)azetidine hydrochloride, 95%, Thermo Scientific Chemicals

81.00€ - 229.14€

Chemical Identifiers

CAS 577777-20-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD12963843,MFCD08061963
InChI Key LHUFQYUQIUJJIB-UHFFFAOYSA-N
Synonym tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-methylaminoazetidine, 3-n-boc-n-methylamino-azetidine, tert-butyl n-azetidin-3-yl-n-methylcarbamate, carbamic acid, 3-azetidinylmethyl-, 1,1-dimethylethyl ester, 3-boc-3-methylamino azetidine hydrochloride, 3-boc-methylamino azetidine, 3-n-boc-methylamino azatidine, 3-boc-3-methylaminoazetidine hcl, tert-butyl azetidin-3-yl-methylcarbamate
PubChem CID 23033645
IUPAC Name tert-butyl N-(azetidin-3-yl)-N-methylcarbamate
SMILES CN(C1CNC1)C(=O)OC(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15462436
SDS View Documents
Thermo Scientific Alfa Aesar
H56060.MD
250 mg
81.00€
250mg
Please to purchase this item. Need a web account? Register with us today!
15442498
SDS View Documents
Thermo Scientific Alfa Aesar
H56060.03
1 g
229.14€
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water. (5 g/100ml)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Boc-3-(methylamino)azetidine hydrochloride, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.