missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

60.70€ - 211.00€

Chemical Identifiers

CAS 202865-68-7
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.50
MDL Number MFCD00143094
InChI Key RSFNYNQGERWCMT-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzylamine hydrochloride, 3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorobenzylamine hcl, 1-3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorophenyl methylamine, chloride, 1-3-bromo-4-fluorophenyl methylamine hydrochloride, benzenemethanamine, 3-bromo-4-fluoro-, hydrochloride, pubchem3226, 3-bromo-4-fluorobenzylaminehydrochloride, acmc-1bczmShow MoreShow Less
PubChem CID 2724901
IUPAC Name (3-bromo-4-fluorophenyl)methanamine;hydrochloride
SMILES Cl.NCC1=CC(Br)=C(F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11499536
SDS View Documents
Thermo Scientific Alfa Aesar
A19280.06
5 g
60.70€
5g
Please to purchase this item. Need a web account? Register with us today!
11409546
SDS View Documents
Thermo Scientific Alfa Aesar
A19280.14
25 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-4-fluorobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.