missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-4-hydroxy-5-methoxycinnamic acid, 97% Product Code.: 11410471

3-Bromo-4-hydroxy-5-methoxycinnamic acid, 97%

Product Code. 11410471
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11410471

Brand: Thermo Scientific Alfa Aesar B20014.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 6948-33-0 | C10H8BrO4 | 272.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6948-33-0
Molecular Formula C10H8BrO4
Molecular Weight (g/mol) 272.08
MDL Number MFCD00017334
InChI Key NGBVBJQUJJKRGF-NSCUHMNNSA-M
Synonym 3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid, 5-bromoferulic acid, 2e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid, 3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid, 3-bromo-4-hydroxy-5-methoxycinnamic acid, 2e-3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid, 2-propenoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl, e-3-3-bromo-4-hydroxy-5-methoxy-phenyl prop-2-enoic acid, e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid
PubChem CID 735789
SMILES COC1=CC(\C=C\C([O-])=O)=CC(Br)=C1O

Specifications

Melting Point 255°C to 257°C
Odor Odorless
Quantity 5 g
IUPAC Name (E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoic acid
Formula Weight 273.09
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxycinnamic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-4-hydroxy-5-methoxycinnamic acid, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.