missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™ Product Code.: 15419065

3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. 15419065
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15419065

Brand: Thermo Scientific Alfa Aesar H35970.MD

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858240-14-8
Molecular Formula C8H6BrClN2O3
Molecular Weight (g/mol) 293.501
MDL Number MFCD22682824
InChI Key RYTIKZUUZLHHDR-UHFFFAOYSA-N
Synonym 3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate
PubChem CID 121235573
IUPAC Name 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
SMILES C1=CC2=NC(=C(N2C=C1Cl)Br)C(=O)O.O

Specifications

Melting Point 216°C to 219°C (decomposition)
Quantity 250 mg
Formula Weight 275.49 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10179583 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.