missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals

Product Code. 11444177
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

Solubility
Slightly soluble in water (3.7 g/L) (25°C)

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 39115-96-3
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007600
InChI Key BNAQRAZIPAHWAR-UHFFFAOYSA-N
Synonym 3-bromobenzhydrazide, m-bromobenzohydrazide, 3-bromobenzoic hydrazide, benzoic acid, 3-bromo-, hydrazide, 3-bromobenzoic acid hydrazide, 3-bromobenzoylhydrazine, benzoylhydrazine, m-bromo, 3-bromo-benzoic acid hydrazide, 3-bromobenzenecarbohydrazide, 3-bromanylbenzohydrazide
PubChem CID 520941
IUPAC Name 3-bromobenzohydrazide
SMILES C1=CC(=CC(=C1)Br)C(=O)NN

Specifications

Melting Point 154°C to 157°C
Quantity 25 g
Beilstein 1945814
Solubility Information Slightly soluble in water (3.7g/L) (25°C)
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.