missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™ Product Code.: 11372669

3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 11372669
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11372669

Brand: Thermo Scientific Alfa Aesar H26711.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2,4-difluorophenylacetic acid is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886761-66-6
Molecular Formula C8H5ClF2O2
Molecular Weight (g/mol) 206.573
MDL Number MFCD06660337
InChI Key HENNVWWQJGUCAH-UHFFFAOYSA-N
Synonym 3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoroShow MoreShow Less
PubChem CID 17750824
IUPAC Name 2-(3-chloro-2,4-difluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1CC(=O)O)F)Cl)F

Specifications

Melting Point 107°C to 110°C
Quantity 1 g
Formula Weight 206.58
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,4-difluorophenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10160020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10184782 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10221791 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10141467 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10165935 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.