missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ Product Code.: 11486236

3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11486236
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11486236

Brand: Thermo Scientific Alfa Aesar B23049.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186517-41-9
Molecular Formula C8H3ClF4O2
Molecular Weight (g/mol) 242.554
MDL Number MFCD01631342
InChI Key PCNHWBAPTNAREE-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-6-trifluoromethyl benzoic acid, benzoic acid,3-chloro-2-fluoro-6-trifluoromethyl, pubchem8541, rarechem al be 0494, acmc-1bx8a, 3-chloro-2-fluoro-6-trifluoromethyl-benzoic acid, benzoic acid, 3-chloro-2-fluoro-6-trifluoromethyl, 3-chloranyl-2-fluoranyl-6-trifluoromethyl benzoic acid, 3-chloro-2-fluoro-6-trifluoromethyl benzoic acid, 97+%
PubChem CID 2773754
IUPAC Name 3-chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1C(F)(F)F)C(=O)O)F)Cl

Specifications

Melting Point 96°C to 98°C
Quantity 5 g
Formula Weight 242.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162339 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10167208 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.