missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-4-methoxybenzylamine hydrochloride, 97%, Thermo Scientific™ Product Code.: 15410699

3-Chloro-4-methoxybenzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 15410699
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15410699

Brand: Thermo Scientific Alfa Aesar H63806.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41965-95-1
Molecular Formula C8H11Cl2NO
Molecular Weight (g/mol) 208.082
MDL Number MFCD01211460
InChI Key IKWWOZCEHOYKAO-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochlorideShow MoreShow Less
PubChem CID 2764287
IUPAC Name (3-chloro-4-methoxyphenyl)methanamine;hydrochloride
SMILES COC1=C(C=C(C=C1)CN)Cl.Cl

Specifications

Melting Point 250°C to 255°C
Quantity 1 g
UN Number UN2811
Formula Weight 208.09
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Chloro-4-methoxybenzylamine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot NumberI23Z001 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-4-methoxybenzylamine hydrochloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.